Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triton™ X-100, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
Physical Form | Viscous Liquid |
CAS Min % | 0.98 |
CAS Max % | 1.0 |
Concentration | 100% |
pH | 6 |
Quantity | 20 L |
Packaging | Cubitainer |
Grade | Laboratory |
Chemical Name or Material | Triton™ X-100 Wetting Agent |
Safety and Handling
ShelfLife | 24 months |
Triton is a trademark of Union Carbide Corporation, a wholly owned subsidiary of The Dow Chemical Company.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction