Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Antibody (165131R), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349670 Supplier Novus Biologicals Supplier No. FAB1751RMFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for CyTOF, Flow Cytometry, Gene Knock-Out.

Specifications

Antigen TrkA
Applications CyTOF, Flow Cytometry, Gene Knock-Out
Classification Monoclonal
Clone 165131R
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession # P04629
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.