Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Antibody (994307), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314189 Supplier Novus Biologicals Supplier No. FAB1752PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TrkA
Applications Flow Cytometry
Classification Monoclonal
Clone 994307
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession # P04629
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.