Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Antibody (MNAC13), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB015440 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015440 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015440 Supplier Novus Biologicals Supplier No. NBP281074AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TrkA
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MNAC13
Conjugate Alexa Fluor 647
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Gene Symbols NTRK1
Host Species Rabbit
Immunogen Human TrkA ECD.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4914
Test Specificity This antibody binds to the extracellular domain of TrkA.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.