Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Antibody (OTI5B6), Novus Biologicals™
SDP

Catalog No. NBP246266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246266 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246266 Supplier Novus Biologicals Supplier No. NBP246266
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

TrkA Monoclonal specifically detects TrkA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TrkA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5B6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Gene Symbols NTRK1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 440-796 of human NTRK1(NP_002520) produced in E.coli.
Molecular Weight of Antigen 87.3 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Neuroscience, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.