Learn More
Description
Specifications
Specifications
| Antigen | TrkA |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | RM423 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:4000, Immunohistochemistry 1:100-1:400 |
| Formulation | PBS, 50% Glycerol, 1% BSA |
| Gene Alias | DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to the C-terminus of human TrkA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
