Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkB Antibody (72509), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB371040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371040 Supplier Novus Biologicals Supplier No. FAB3971D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TrkB Monoclonal specifically detects TrkB in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TrkB
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 72509
Conjugate DyLight 350
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TrkB, Cys32-His430, Accession # Q16620
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroendocrinology, Oncogenes, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4915
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.