Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkB Antibody (8D2E8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252523 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252523 Supplier Novus Biologicals Supplier No. NBP252523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TrkB Monoclonal specifically detects TrkB in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.

Specifications

Antigen TrkB
Applications Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 8D2E8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B
Gene Symbols NTRK2
Host Species Mouse
Immunogen Purified recombinant fragment of human NTRK2 (AA: 207-339) expressed in E. Coli.
Molecular Weight of Antigen 92 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4915
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.