Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkB Rabbit anti-Human, Mouse, Rat, Clone: S04-6B1, Novus Biologicals™
SDP

Catalog No. NB159276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159276 25 μg
NB159275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159276 Supplier Novus Biologicals Supplier No. NBP31490225UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TrkB Monoclonal antibody specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TrkB
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S04-6B1
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 2.5 ug/mL, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Tyr705 of human TrkB (Uniprot#: Q16620)
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.