Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkC Antibody (75213), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB296398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296398 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296398 Supplier Novus Biologicals Supplier No. FAB3731A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TrkC Monoclonal antibody specifically detects TrkC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen TrkC
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 75213
Conjugate APC
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TrkC, Cys32-Asp428, Accession # Q16288
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4916
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.