Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkC Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB004913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004913 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004913 Supplier Novus Biologicals Supplier No. NBP215185H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TrkC Polyclonal specifically detects TrkC in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TrkC
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C
Gene Symbols NTRK3
Host Species Rabbit
Immunogen A synthetic peptide made to the extracellular domain of the human TrkC protein (within residues 300-400). [UniProt Q16288].
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.