Learn More
Description
Specifications
Specifications
| Antigen | TrkC |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.26mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:200 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Accession No. | Q16288, Q6VNS1, Q03351 |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C |
| Gene Symbols | NTRK3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
