Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkC Antibody, Novus Biologicals™
SDP

Catalog No. NB10098829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10098829 0.1 mL
NB017953 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10098829 Supplier Novus Biologicals Supplier No. NB10098829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TrkC Polyclonal specifically detects TrkC in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TrkC
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q16288, Q6VNS1, Q03351
Gene Alias EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C
Gene Symbols NTRK3
Host Species Rabbit
Immunogen A synthetic peptide from the extracellular domain of rat TrkC conjugated to blue carrier protein was used as the antigen. The peptide is homologous in mouse and human.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4916
Test Specificity Human, mouse and rat. Immunogen has 100% homology to pig.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.