Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT12 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357872 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357872 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357872 Supplier Novus Biologicals Supplier No. NBP176583MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRMT12 Polyclonal antibody specifically detects TRMT12 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen TRMT12
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Alpha-amino-alpha-carboxypropyl transferase TYW2, EC 2.1.1, EC 2.1.1.32, EC 2.5.1.-, FLJ20772, homolog of yeast tRNA methyltransferase, Trm12, tRNA methyltranferase 12 homolog, tRNA methyltranferase 12 homolog (S. cerevisiae), tRNA methyltransferase 12 homolog (S. cerevisiae), tRNA-yW synthesizing protein 2, tRNA-yW-synthesizing protein 2, TYW2tRNA wybutosine-synthesizing protein 2 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TYW2. The immunogen is located within the first 50 amino acids of TYW2. Amino Acid Squence: TQRYREYLQRQKLFDTQHR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55039
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.