Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT61A Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169971 100 μg
NB169972 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB169971 Supplier Novus Biologicals Supplier No. NBP317448100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRMT61A Polyclonal antibody specifically detects TRMT61A in Human samples. It is validated for Immunofluorescence

Specifications

Antigen TRMT61A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C14orf172, chromosome 14 open reading frame 172, EC 2.1.1.36, FLJ40452, GCD14, Gcd14p, hTRM61, TRM61, tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRM61, tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61A, tRNA methyltransferase 61 homolog A (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM61, tRNA(m1A58)-methyltransferase subunit TRMT61A, tRNA(m1A58)MTase subunit TRM61, tRNA(m1A58)MTase subunit TRMT61A
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: GSGSVSHAIIRTIAPTGHLHTVEFHQQRAEKAREEFQEHRVGRWVTVRTQDVCRSGFGVSHVADAVFPDIPSPWEAVGHAWDALKVEGGRFCSFSPCIEQVQRTCQALAARGFSELSTLEVLPQVYNVRTVSLPPPDLGTGTDGPA
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115708
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.