Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROP-2 Antibody (TACSTD2/2152), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB008798 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008798 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008798 Supplier Novus Biologicals Supplier No. NBP275746AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TROP-2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TACSTD2/2152
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2
Gene Symbols TACSTD2
Host Species Mouse
Immunogen Recombinant fragment of human TROP-2 protein (around aa 31-274) (exact sequence is proprietary) (Uniprot: P09758 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Test Specificity TROP-2 is a cell surface glycoprotein receptor. It is a single pass type I membrane protein containing one thyroglobulin type-1 domain, an epidermal growth factor-like repeat, a phosphatidylinositol binding site and tyrosine phosphorylation sites near the C-terminus. It plays a role in transducing intracellular calcium signals. It is expressed in trophoblast cells, cornea and multi-stratified epithelia. It is also highly expressed in several types of tumors and is involved in regulating the growth of carcinoma cells.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.