Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trophinin Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB343367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343367 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343367 Supplier Novus Biologicals Supplier No. NBP241320MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trophinin Polyclonal antibody specifically detects Trophinin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Trophinin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias KIAA1114, MAGE Superfamily Protein, MAGE-d3, MAGED3, MAGE-D3 Antigen, magphinin, Melanoma Antigen, Family D, 3, trophinin
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the amino terminus of human Trophinin. The immunogen is located within amino acids 390 - 440 of Trophinin. Amino Acid Squence: EPTTRTRGKRNRKSKH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7216
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.