Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tropisetron hydrochloride
GREENER_CHOICE

Catalog No. p-4784726
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent 5-HT3 antagonist; also α7 nAChR partial agonist

Potent, orally active 5-HT3 receptor antagonist. Antiemetic. Also partial agonist of α7 nAChR. Activates α7Q79G-GlyR chimeric ion channels (EC50 = 38 nM)

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2

Specifications

Quantity 10 mg
Formula Weight 320.82
Purity >99%
Molecular Weight (g/mol) 320.817
Chemical Name or Material Tropisetron hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.