Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin-1 Rabbit anti-Mouse, Rat, Clone: 5Q2H6, Novus Biologicals™
SDP

Catalog No. NB168469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168469 100 μg
NB168470 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168469 Supplier Novus Biologicals Supplier No. NBP316689100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Tropomyosin-1 Monoclonal antibody specifically detects Tropomyosin-1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Tropomyosin-1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5Q2H6
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias alpha tropomyosin, alpha-tropomyosin, C15orf13, cardiomyopathy, hypertrophic 3, chromosome 15 open reading frame 13, CMD1Y, CMH3, HTM-alpha, sarcomeric tropomyosin kappa, TMSA, tropomyosin 1 (alpha), tropomyosin 1 (alpha) isoform 1, tropomyosin 1 (alpha) isoform 2, tropomyosin 1 (alpha) isoform 3, tropomyosin 1 (alpha) isoform 4, tropomyosin 1 (alpha) isoform 5, tropomyosin 1 (alpha) isoform 6, tropomyosin 1 (alpha) isoform 7, tropomyosin alpha-1 chain, Tropomyosin-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 185-284 of human Tropomyosin-1 (P09493).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7168
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.