Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin 3 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 2G10.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT133525UG
Description
Tropomyosin 3 Monoclonal antibody specifically detects Tropomyosin 3 in Human,Mouse,Rat samples. It is validated for Immunofluorescence, Western Blotting.
Specifications
Tropomyosin 3 | |
Monoclonal | |
Unconjugated | |
Gamma-tropomyosin;Tropomyosin-3;Tropomyosin-5;TPM3;Tm3 | |
Mouse | |
∼30 kDa observed; 29.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunofluorescence, Western Blot | |
2G10.2 | |
NP_001288215 | |
Tpm3, Tpm-5, Tpm5 | |
Diphtheria Toxoid Carrier Protein-conjugated linear peptide corresponding to 27 amino acids from the C-terminal region of rat Tropomyosin alpha-3 chain, isoform 1. | |
Protein G Purified | |
Cell Structure | |
Clone 2G10.2 is a mouse monoclonal antibody that specifically detects Tropomyosin alpha-3 chain. It targets an epitope within 27 amino acids from the C-terminal region. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction