Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tropomyosin 3 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 2G10.2, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | Tropomyosin 3 |
---|---|
Clone | 2G10.2 |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT13352MI
|
MilliporeSigma
MABT133525UG |
25 μg |
Each for $189.29
|
|
|||||
MABT13351MI
|
MilliporeSigma
MABT1335100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
Tropomyosin 3 Monoclonal antibody specifically detects Tropomyosin 3 in Human,Mouse,Rat samples. It is validated for Immunofluorescence, Western Blotting.Specifications
Tropomyosin 3 | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Gamma-tropomyosin;Tropomyosin-3;Tropomyosin-5;TPM3;Tm3 | |
Diphtheria Toxoid Carrier Protein-conjugated linear peptide corresponding to 27 amino acids from the C-terminal region of rat Tropomyosin alpha-3 chain, isoform 1. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼30 kDa observed; 29.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
2G10.2 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_001288215 | |
Tpm3, Tpm-5, Tpm5 | |
IgG2b κ | |
Protein G Purified | |
Clone 2G10.2 is a mouse monoclonal antibody that specifically detects Tropomyosin alpha-3 chain. It targets an epitope within 27 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title