Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin C (cardiac) Antibody (7B9cc), Novus Biologicals™
SDP

Catalog No. NB001508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001508 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001508 Supplier Novus Biologicals Supplier No. NBP278721
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin C (cardiac) Monoclonal specifically detects Troponin C (cardiac) in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Troponin C (cardiac)
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7B9cc
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow
Gene Symbols TNNC1
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human cardiac troponin complex.
Purification Method Protein A purified
Quantity 0.2 mg
Research Discipline Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7134
Test Specificity No cross-reactivity with cardiac Troponin T and Troponin I. Also recognizes Troponin C in binary complex with cardiac Troponin I.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.