Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin C (cardiac) Antibody (OTI5H3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274638G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274638G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274638G Supplier Novus Biologicals Supplier No. NBP274638G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin C (cardiac) Monoclonal specifically detects Troponin C (cardiac) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Troponin C (cardiac)
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI5H3
Conjugate DyLight 488
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow
Gene Symbols TNNC1
Host Species Mouse
Immunogen Full length recombinant protein of human Troponin C (cardiac) (NP_003271) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7134.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.