Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 1 (slow skeletal) Antibody (36E7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB170843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
50 ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170843 50 ul
NBP222621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170843 Supplier Novus Biologicals Supplier No. NBP22262150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Troponin I Type 1 (slow skeletal) Monoclonal specifically detects Troponin I Type 1 (slow skeletal) in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Troponin I Type 1 (slow skeletal)
Applications Western Blot
Classification Monoclonal
Clone 360000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA 1:100-1:2000
Gene Accession No. NP_003272
Gene Alias DKFZp451O223, slow-twitch isoform, SSTNI, TNN1, troponin I type 1 (skeletal, slow), troponin I, skeletal, slow, troponin I, slow skeletal muscle
Gene Symbols TNNI1
Host Species Mouse
Immunogen Recombinant human TNNI1 (1-187aa) purified from E. coli
Purification Method Protein A purified
Quantity 50 ul
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7135
Test Specificity Anti-human TNNI1 mAb, clone 36E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TNNI1 protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.