Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 1 (slow skeletal) Antibody (36E7), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309997 Supplier Novus Biologicals Supplier No. NBP222621MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 1 (slow skeletal) Monoclonal antibody specifically detects Troponin I Type 1 (slow skeletal) in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Troponin I Type 1 (slow skeletal)
Applications Western Blot, ELISA
Classification Monoclonal
Clone 36E7
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp451O223, slow-twitch isoform, SSTNI, TNN1, troponin I type 1 (skeletal, slow), troponin I, skeletal, slow, troponin I, slow skeletal muscle
Host Species Mouse
Immunogen Recombinant human Troponin I Type 1 (slow skeletal) (1-187aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7135
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.