Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 1 (slow skeletal) Antibody (OTI8H8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274639A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274639A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274639A70 Supplier Novus Biologicals Supplier No. NBP274639AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 1 (slow skeletal) Monoclonal specifically detects Troponin I Type 1 (slow skeletal) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Troponin I Type 1 (slow skeletal)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8H8
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp451O223, slow-twitch isoform, SSTNI, TNN1, troponin I type 1 (skeletal, slow), troponin I, skeletal, slow, troponin I, slow skeletal muscle
Gene Symbols TNNI1
Host Species Mouse
Immunogen Full length recombinant protein of human Troponin I Type 1 (slow skeletal) (NP_003272) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7135
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.