Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I type 2 (fast skeletal) Antibody (OTI3A11), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274640B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274640B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274640B Supplier Novus Biologicals Supplier No. NBP274640B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I type 2 (fast skeletal) Monoclonal specifically detects Troponin I type 2 (fast skeletal) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Troponin I type 2 (fast skeletal)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3A11
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias skeletal, fast, troponin I type 2 (skeletal, fast), troponin I, fast skeletal muscle, Troponin I, fast-twitch isoform, troponin I, fast-twitch skeletal muscle isoform
Gene Symbols TNNI2
Host Species Mouse
Immunogen Full length recombinant protein of human Troponin I type 2 (fast skeletal) (NP_003273) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7136.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.