Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 3 (cardiac) Antibody (16A11cc), Biotin, Novus Biologicals™
SDP

Catalog No. NB001750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001750 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001750 Supplier Novus Biologicals Supplier No. NB10073082B200UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for ELISA, Immunoaffinity Purification, Immunoassay, Immunohistochemistry, Immunoprecipitation, Sandwich ELISA, Western Blot.

Specifications

Antigen Troponin I Type 3 (cardiac)
Applications ELISA, Immunoaffinity Chromatography, Immunoassay, Immunohistochemistry, Immunoprecipitation, Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 16A11
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Gene Symbols TNNI3
Host Species Mouse
Immunogen free human cTnI, native human cTn complex, or human cTnI peptides. a.a.r. 86-90.
Quantity 200 μL
Research Discipline Apoptosis, Cell Biology, Cellular Markers, Phospho Specific, Stem Cell Markers
Gene ID (Entrez) 7137
Test Specificity This is reactive against troponin.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.