Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 3 (cardiac) Antibody (1E10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP235337SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP235337SS 0.025 mg
NBP235337 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP235337SS Supplier Novus Biologicals Supplier No. NBP235337SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Troponin I Type 3 (cardiac)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10000000000
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. P19429
Gene Alias Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Gene Symbols TNNI3
Host Species Mouse
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7137
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.