Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 3 (cardiac) Antibody (1E10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB337047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337047 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337047 Supplier Novus Biologicals Supplier No. NBP235337MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Troponin I Type 3 (cardiac)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Host Species Mouse
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cellular Markers, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7137
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.