Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Type 3 (cardiac) Antibody (MF4cc), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB12010239Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010239Q 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010239Q Supplier Novus Biologicals Supplier No. NB12010239IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human, Rat, Porcine, Bovine, Canine, Feline, Goat, Fish (Negative), Rabbit (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Troponin I Type 3 (cardiac)
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MF4cc
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Gene Symbols TNNI3
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with free human cTnI.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7137
Test Specificity No cross-reaction with skeletal TnT
Target Species Human, Rat, Pig, Bovine, Canine, Feline, Goat, Fish (Negative), Rabbit (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.