Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T Type 2 (cardiac) Antibody (BGN/1288/44), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1066490A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066490A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066490A30 Supplier Novus Biologicals Supplier No. NB10066490AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Troponin T Type 2 (cardiac) Monoclonal specifically detects Troponin T Type 2 (cardiac) in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Troponin T Type 2 (cardiac)
Applications Western Blot, ELISA
Classification Monoclonal
Clone BGN/1288/44
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle
Gene Symbols TNNT2
Host Species Mouse
Immunogen HMW conjugate
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7139.0
Test Specificity NB100-66490 recognises Troponin T Type 2 (cardiac) isoform 1. In Western blotting, this reacts with reduced purified cardiac isoform, but does not react with the skeletal isoform.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.