Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T Type 2 (cardiac) Antibody (CDC4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20294A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20294A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20294A94 Supplier Novus Biologicals Supplier No. NBP250294AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Troponin T Type 2 (cardiac) Monoclonal specifically detects Troponin T Type 2 (cardiac) in Rat, Chicken samples. It is validated for Western Blot.

Specifications

Antigen Troponin T Type 2 (cardiac)
Applications Western Blot
Classification Monoclonal
Clone CDC4
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle
Gene Symbols TNNT2
Host Species Mouse
Immunogen Purified chicken Troponin T Type 2 (cardiac)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7139
Test Specificity This particular antibody recognises slow skeletal and cardiac muscle Troponin T.
Target Species Rat, Chicken
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.