Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T Type 2 (cardiac) Antibody (CDC4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309628 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309628 Supplier Novus Biologicals Supplier No. NBP250294MFV500

Mouse Monoclonal Antibody

Troponin T Type 2 (cardiac) Monoclonal antibody specifically detects Troponin T Type 2 (cardiac) in Rat, Chicken samples. It is validated for Western Blot

Specifications

Antigen Troponin T Type 2 (cardiac)
Applications Western Blot
Classification Monoclonal
Clone CDC4
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle
Host Species Mouse
Immunogen Purified chicken Troponin T Type 2 (cardiac)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7139
Target Species Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.