Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T Type 3 (fast skeletal) Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB331085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331085 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331085 Supplier Novus Biologicals Supplier No. NBP297579PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Troponin T Type 3 (fast skeletal) Polyclonal antibody specifically detects Troponin T Type 3 (fast skeletal) in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Troponin T Type 3 (fast skeletal)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Beta-TnTF, fast skeletal muscle, TnTf, troponin T type 3 (skeletal, fast), troponin T3, skeletal, fast, troponin-T3, skeletal, fast
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Troponin T Type 3 (fast skeletal) fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7140
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.