Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROVE2 Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB104485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104485 Supplier Novus Biologicals Supplier No. NBP298552IR

Rabbit Polyclonal Antibody

TROVE2 Polyclonal specifically detects TROVE2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TROVE2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias 60 kDa ribonucleoprotein Ro, 60 kDa Ro protein, 60 kDa SS-A/Ro ribonucleoprotein, Ro 60 kDa autoantigen, RO60gastric cancer multi-drug resistance protein, RORNP, Sjoegren syndrome antigen A2, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A2 (60kDa, ribonucleoprotein autoantigen SS-A/Ro), SS-A, SSA2Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro), TROVE domain family member 2, TROVE domain family, member 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TROVE2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6738
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.