Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROY/TNFRSF19 Antibody (933202), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349836 Supplier Novus Biologicals Supplier No. FAB15481MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TROY/TNFRSF19
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 933202
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19, Glu30-Leu170, Accession # Q9NS68
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55504
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.