Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROY/TNFRSF19 Rabbit anti-Mouse, Clone: 104, Novus Biologicals™
SDP

Catalog No. NB022354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022354 50 μg
NB022353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022354 Supplier Novus Biologicals Supplier No. NBP29045050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for Sandwich ELISA Capture.

Specifications

Antigen TROY/TNFRSF19
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate Unconjugated
Dilution Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Host Species Rabbit
Immunogen Recombinant Mouse TROY/TNFRSF19 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55504
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.