Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRP7 Antibody (N64A/36), FITC, Novus Biologicals™
SDP

Catalog No. NBP148318F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP148318F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP148318F Supplier Novus Biologicals Supplier No. NBP148318F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRP7 Monoclonal specifically detects TRP7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TRP7
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S64A-36
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9HCX4, NP_065122.1
Gene Alias hTRP7, KNP3, likley ortholog of mouse transient receptor potential cation channel, subfamilyC, member 7, putative capacitative calcium channel, short transient receptor potential channel 7, transient receptor potential cation channel, subfamily C, member 7, transient receptor potential-related channel 7, a novel putative Ca2+ channelprotein10TRPM2, Transient receptor protein 7, TRP7, TRP-7, TrpC7
Gene Symbols TRPC7
Host Species Mouse
Immunogen Synthetic peptide amino acids 845-862 of human TRPC7
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57113
Test Specificity Detects approx 100kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.