Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRP7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144058 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144058 Supplier Novus Biologicals Supplier No. NBP144058
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

TRP7 Polyclonal specifically detects TRP7 in Chicken samples. It is validated for Western Blot, Immunoprecipitation, Peptide ELISA.

Specifications

Antigen TRP7
Applications Immunoprecipitation, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 1:50, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065122.1
Gene Alias hTRP7, KNP3, likley ortholog of mouse transient receptor potential cation channel, subfamilyC, member 7, putative capacitative calcium channel, short transient receptor potential channel 7, transient receptor potential cation channel, subfamily C, member 7, transient receptor potential-related channel 7, a novel putative Ca2+ channelprotein10TRPM2, Transient receptor protein 7, TRP7, TRP-7, TrpC7
Gene Symbols TRPC7
Host Species Goat
Immunogen Peptide with sequence KLCKSKAKSCEND corresponding to internal region according to NP_001161049.1, NP_001161048.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57113
Test Specificity This antibody is expected to cross-react with isoform 1, 2 and 3 (NP_065122.1; NP_001161049.1; NP_001161048.1).
Target Species Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.