Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB184040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184040 Supplier Novus Biologicals Supplier No. NBP282043AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRPC3 Polyclonal antibody specifically detects TRPC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TRPC3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias hTrp3, hTrp-3, short transient receptor potential channel 3, transient receptor potential cation channel, subfamily C, member 3, transient receptor potential channel 3, Transient receptor protein 3, TRP3, TRP-3, TrpC3
Host Species Rabbit
Immunogen TRPC3 antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human TRPC3. The immunogen is located within amino acids 710 - 760 of TRPC3. Amino Acid Squence: RRRRLQKDIEMGMGNSKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7222
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.