Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC4AP Mouse anti-Human, Clone: 2C10B1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TRPC4AP |
---|---|
Clone | 2C10B1 |
Concentration | 0.96 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6574
![]() ![]() |
Proteintech Group Inc
661381IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.Specifications
TRPC4AP | |
0.96 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C20orf188, Protein TAP1, Protein TRUSS, Trp4 associated protein, Trpc4 associated protein, TRPC4AP, TRRP4AP, TRUSS | |
TRPC4AP | |
IgG3 | |
Protein A | |
Antibody |
2C10B1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8TEL6 | |
26133 | |
TRPC4AP Fusion Protein Ag18772 | |
Primary | |
-20°C | |
TRPC4AP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title