Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC5 Antibody (N67/15), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB362589 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362589 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362589 Supplier Novus Biologicals Supplier No. NBP212919AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRPC5 Monoclonal antibody specifically detects TRPC5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray

Specifications

Antigen TRPC5
Applications Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Microarray
Classification Monoclonal
Clone N67/15
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
Formulation 50mM Sodium Borate
Gene Alias hTRP5, hTRP-5, short transient receptor potential channel 5, transient receptor potential cation channel, subfamily C, member 5, Transient receptor protein 5, TRP-5, TRP5transient receptor potential channel 5, TrpC5
Host Species Mouse
Immunogen Synthetic peptide amino acids 827-845 of human TrpC5 (also known as short transient receptor potential channel 5, and Htrp5)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7224
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.