Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC6 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356159 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356159 Supplier Novus Biologicals Supplier No. NBP177260PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPC6 Polyclonal antibody specifically detects TRPC6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TRPC6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ11098, FSGS2, short transient receptor potential channel 6, transient receptor potential cation channel, subfamily C, member 6, Transient receptor protein 6, TRP-6, TRP6FLJ14863, TrpC6
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human TRPC6. The immunogen is located within the last 50 amino acids of TRPC6. Amino Acid Squence: LEEKSQNTEDLAEL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7225
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.