Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018018 0.025 mL
NB11074925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018018 Supplier Novus Biologicals Supplier No. NB110749250.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPM1 Polyclonal specifically detects TRPM1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TRPM1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000
Formulation Whole antisera with No Preservative
Gene Accession No. Q2TV84
Gene Alias CSNB1C, Long transient receptor potential channel 1, LTrpC1, LTRPC1transient receptor potential cation channel subfamily M member 1, melastatin 1, melastatin-1, MLSN, MLSN1transient receptor potential melastatin family, transient receptor potential cation channel, subfamily M, member 1
Gene Symbols TRPM1
Host Species Rabbit
Immunogen A synthetic peptide from mouse TRPM1 conjugated to immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4308
Test Specificity The protein encoded by this gene is similar to the transient receptor potential (TRP) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration. Add glycerol (1:1) for additional stability.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.