Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB11081601Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11081601Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11081601Y Supplier Novus Biologicals Supplier No. NB11081601AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPM2 Polyclonal specifically detects TRPM2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen TRPM2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7
Gene Symbols TRPM2
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot# Q5G856]
Molecular Weight of Antigen 181 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7226
Test Specificity This is specific for isoform 1, or the long isoform (TRPM2-L), of TRPM2.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.