Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM5 Antibody, Novus Biologicals™
SDP

Catalog No. NB036116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036116 Supplier Novus Biologicals Supplier No. NBP288494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPM5 Polyclonal specifically detects TRPM5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TRPM5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose
Gene Alias Long transient receptor potential channel 5, LTrpC5, LTrpC-5, LTRPC5transient receptor potential cation channel subfamily M member 5, MLSN1- and TRP-related gene 1 protein, MTR1MLSN1 and TRP-related, transient receptor potential cation channel, subfamily M, member 5
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of human TRPM5
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29850
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.