Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM7 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018402 0.1 mg
NBP120224 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018402 Supplier Novus Biologicals Supplier No. NBP1202240.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TRPM7 Polyclonal specifically detects TRPM7 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TRPM7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 30 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml
Formulation PBS with No Preservative
Gene Accession No. Q96QT4
Gene Alias CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK
Gene Symbols TRPM7
Host Species Rabbit
Immunogen A synthetic peptide from the 4th cytoplasmic loop of human TRPM7 (LTRPC7) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54822
Reconstitution Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability.
Target Species Rat, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.