Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500243SS
Description
TRPM7 Polyclonal specifically detects TRPM7 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TRPM7 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation 5 ul | |
CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
NB 500-243 recognizes overexpressed human TRPM7. It does not appear to recognize overexpressed mouse TRPM7. It does not work on endogenous samples tested for western analysis (human, rat and mouse cell lines. Other species have not been tested. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
TRPM7 | |
A synthetic peptide made to the C-terminal region of rat TRPM7. [UniProt# Q925B3] | |
0.025 mL | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
54822 | |
Human, Mouse (Negative) | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction