Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM7 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen TRPM7
Dilution Western Blot 1:1000, Immunoprecipitation 5 ul
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB500243SS
SDP
View Documents
Novus Biologicals
NB500243SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NB500243
SDP
View Documents
Novus Biologicals
NB500243
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

TRPM7 Polyclonal specifically detects TRPM7 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TRPM7
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Whole antisera with 0.05% Sodium Azide
54822
A synthetic peptide made to the C-terminal region of rat TRPM7. [UniProt# Q925B3]
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot 1:1000, Immunoprecipitation 5 ul
Polyclonal
Antisera
RUO
Human, Mouse (Negative)
CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK
TRPM7
IgG
Unpurified
NB 500-243 recognizes overexpressed human TRPM7. It does not appear to recognize overexpressed mouse TRPM7. It does not work on endogenous samples tested for western analysis (human, rat and mouse cell lines. Other species have not been tested.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.